Product Details

SNP ID
rs4791658
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15981478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATAAGAAGGAAGTGGGGGGGGG[A/G]AATCTCTCATCTACATTTTATTGTG
Phenotype
MIM: 600446 MIM: 614535
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADORA2B PubMed Links
Additional Information
For this assay, SNP(s) [rs151298772] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ADORA2B
Gene Name
adenosine A2b receptor
There are no transcripts associated with this gene.

Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 Intron NP_001036162.1
NM_001042698.1 Intron NP_001036163.1

View Full Product Details