Product Details

SNP ID
rs10454027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99607122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGTTCGTTTAACACAGCGGTTCC[C/T]AAACGGTTTGGTCTCAGGACCCGTT
Phenotype
MIM: 609686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLYBL PubMed Links

Gene Details

Gene
CLYBL
Gene Name
citrate lyase beta like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206808.3 458 Intron NP_996531.1
XM_005254030.2 458 Intron XP_005254087.1
XM_005254031.2 458 Intron XP_005254088.1
XM_006719915.3 458 Intron XP_006719978.1
XM_011521051.2 458 Intron XP_011519353.1
XM_011521052.2 458 Intron XP_011519354.1
XM_017020402.1 458 Intron XP_016875891.1
XM_017020403.1 458 UTR 5 XP_016875892.1
XM_017020404.1 458 Intron XP_016875893.1

View Full Product Details