Product Details
- SNP ID
-
rs7276755
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:29202215 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCTGCTTTCAAGATTCTTGCTTT[C/G]TCTTTGTCTTTTTACAGTCAGATTA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00189
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139555705] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00189
- Gene Name
- long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.
View Full Product Details