Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321967.1 | 2859 | UTR 3 | NP_001308896.1 | ||
NM_001321968.1 | 2859 | UTR 3 | NP_001308897.1 | ||
NM_001321969.1 | 2859 | UTR 3 | NP_001308898.1 | ||
NM_032810.3 | 2859 | UTR 3 | NP_116199.2 | ||
XM_005270252.4 | 2859 | UTR 3 | XP_005270309.1 | ||
XM_011540302.1 | 2859 | UTR 3 | XP_011538604.1 | ||
XM_011540303.2 | 2859 | Intron | XP_011538605.1 | ||
XM_017016847.1 | 2859 | UTR 3 | XP_016872336.1 | ||
XM_017016848.1 | 2859 | UTR 3 | XP_016872337.1 | ||
XM_017016849.1 | 2859 | UTR 3 | XP_016872338.1 | ||
XM_017016850.1 | 2859 | UTR 3 | XP_016872339.1 | ||
XM_017016851.1 | 2859 | Intron | XP_016872340.1 |