Product Details

SNP ID
rs12146325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:126428870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCTACGAATCAGGTCTTTAATGG[G/T]ACGGTCTCCCTTTTCCTCATTTTCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf90 PubMed Links

Gene Details

Gene
C10orf90
Gene Name
chromosome 10 open reading frame 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004298.2 Intron NP_001004298.2
XM_005269491.1 Intron XP_005269548.1
XM_005269492.2 Intron XP_005269549.1
XM_011539216.1 Intron XP_011537518.1
XM_011539217.2 Intron XP_011537519.1
XM_011539220.2 Intron XP_011537522.1
XM_011539222.2 Intron XP_011537524.1
XM_011539223.2 Intron XP_011537525.1
XM_011539224.2 Intron XP_011537526.1
XM_011539226.1 Intron XP_011537528.1
XM_011539227.2 Intron XP_011537529.1
XM_011539230.2 Intron XP_011537532.1
XM_011539231.1 Intron XP_011537533.1
XM_011539232.1 Intron XP_011537534.1
XM_011539233.1 Intron XP_011537535.1
XM_017015634.1 Intron XP_016871123.1
XM_017015635.1 Intron XP_016871124.1
Gene
LINC00601
Gene Name
long intergenic non-protein coding RNA 601
There are no transcripts associated with this gene.

Gene
LOC728158
Gene Name
hCG2044975
There are no transcripts associated with this gene.

View Full Product Details