Product Details

SNP ID
rs6482850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:131105576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGACTGGGAAATGTTCTGGAGGC[A/G]GGAGGTGATGGAATACTCGTTCATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCERG1L PubMed Links

Gene Details

Gene
TCERG1L
Gene Name
transcription elongation regulator 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174937.3 Intron NP_777597.2
XM_011539577.1 Intron XP_011537879.1
XM_011539578.2 Intron XP_011537880.1
Gene
TCERG1L-AS1
Gene Name
TCERG1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details