Product Details
- SNP ID
-
rs9495317
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:139033001 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAAGTGAAATGAAGAGGGGCACC[C/G]GGTGCTATGGGAACACACAGTGAGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ABRACL
PubMed Links
Gene Details
- Gene
- ABRACL
- Gene Name
- ABRA C-terminal like
View Full Product Details