Product Details

SNP ID
rs4464748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:78938261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGTCTAATAATGTCCAAATAA[C/G]GTCATAAATATACAGTGTCTGTAGA
Phenotype
MIM: 615375 MIM: 612870
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRAK1BP1 PubMed Links

Gene Details

Gene
IRAK1BP1
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010844.3 Intron NP_001010844.1
Gene
PHIP
Gene Name
pleckstrin homology domain interacting protein
There are no transcripts associated with this gene.

View Full Product Details