Product Details

SNP ID
rs3108668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38744908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCAAAAATCTAGTTAGAAACAT[G/T]TTATTTAAATGTGCCAAATAAAAAC
Phenotype
MIM: 600572
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SRSF7 PubMed Links

Gene Details

Gene
SRSF7
Gene Name
serine and arginine rich splicing factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031684.2 1002 UTR 3 NP_001026854.1
NM_001195446.1 1002 UTR 3 NP_001182375.1
XM_005264484.2 1002 UTR 3 XP_005264541.1
XM_005264485.2 1002 UTR 3 XP_005264542.1
XM_011533032.2 1002 UTR 3 XP_011531334.1

View Full Product Details