Product Details
- SNP ID
-
hCV27456837
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:155225058 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTATCTTTAAGTATTTAATGTATAG[C/T]TTTATCTTTTGGTATTTGTTAATTT
- Phenotype
-
MIM: 300133
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VBP1
PubMed Links
Gene Details
- Gene
- VBP1
- Gene Name
- VHL binding protein 1
There are no transcripts associated with this gene.
View Full Product Details