Product Details

SNP ID
rs3130602
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33004430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACATTTGTGTTGAGATCTTGATT[C/T]CTGCTCTAACTACACAGGGCTGGGA
Phenotype
MIM: 142930
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DOA PubMed Links

Gene Details

Gene
HLA-DOA
Gene Name
major histocompatibility complex, class II, DO alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002119.3 3237 UTR 3 NP_002110.1

View Full Product Details