Product Details

SNP ID
rs3176988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42697397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCCACCTGTGTGGGTGTCCCT[G/T]GGCCCACCAAGAGCTGAGGCCTGGT
Phenotype
MIM: 179605 MIM: 609134
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRPH2 PubMed Links

Gene Details

Gene
PRPH2
Gene Name
peripherin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000322.4 2225 UTR 3 NP_000313.2
Gene
UBR2
Gene Name
ubiquitin protein ligase E3 component n-recognin 2
There are no transcripts associated with this gene.

View Full Product Details