Product Details

SNP ID
rs3179087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18531770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGAGTCCGTGGAAACTGCAGGG[G/T]GGGATGCTTTATTTCACTGTGGCGG
Phenotype
MIM: 600284 MIM: 604840
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELL PubMed Links

Gene Details

Gene
ELL
Gene Name
elongation factor for RNA polymerase II
There are no transcripts associated with this gene.

Gene
FKBP8
Gene Name
FK506 binding protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308373.1 1835 UTR 3 NP_001295302.1
NM_012181.4 1835 UTR 3 NP_036313.3
XM_011527863.1 1835 UTR 3 XP_011526165.1
XM_011527864.1 1835 UTR 3 XP_011526166.1

View Full Product Details