Product Details
- SNP ID
-
rs3184994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56270264 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGTCGGGCAGCCACCAAGTTTGGT[C/T]CAGAAACAGCCATCCCCCGTGAACT
- Phenotype
-
MIM: 118950
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COQ10A
PubMed Links
Gene Details
- Gene
- COQ10A
- Gene Name
- coenzyme Q10A
- Gene
- CS
- Gene Name
- citrate synthase
There are no transcripts associated with this gene.
View Full Product Details