Product Details
- SNP ID
-
rs3191540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:89162724 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCCTCTTCCGTGTCGCTGGGGCTC[T/G]CGTGGCTCCGGAAGCTCCCCTCGCT
- Phenotype
-
MIM: 602628
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOXN3
PubMed Links
Gene Details
- Gene
- FOXN3
- Gene Name
- forkhead box N3
View Full Product Details