Product Details
- SNP ID
-
rs3177428
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:91000533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCAGGGGCACATAAGCACCACCG[G/A]AGACGTAAGCCATGTCTCGGGGCAC
- Phenotype
-
MIM: 603484
MIM: 608552
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11073964] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRC1
- Gene Name
- protein regulator of cytokinesis 1
There are no transcripts associated with this gene.
- Gene
- VPS33B
- Gene Name
- VPS33B, late endosome and lysosome associated
View Full Product Details