Product Details

SNP ID
rs3193941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72783821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTACCTACAATGCAGCAGACAG[G/T]AATAAATTCCCCAAGTGAGATAAAG
Phenotype
MIM: 104155
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFHX3 PubMed Links

Gene Details

Gene
ZFHX3
Gene Name
zinc finger homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164766.1 11842 UTR 3 NP_001158238.1
NM_006885.3 11842 UTR 3 NP_008816.3
XM_005255957.3 11842 UTR 3 XP_005256014.1
XM_017023251.1 11842 UTR 3 XP_016878740.1

View Full Product Details