Product Details

SNP ID
rs3210562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90998788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGTACTTCATAAACAGAAAAGA[A/G]AAATATCCTGGGAGCAGGAAGTGAA
Phenotype
MIM: 603484 MIM: 608552
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRC1 PubMed Links

Gene Details

Gene
PRC1
Gene Name
protein regulator of cytokinesis 1
There are no transcripts associated with this gene.

Gene
VPS33B
Gene Name
VPS33B, late endosome and lysosome associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289148.1 2332 UTR 3 NP_001276077.1
NM_001289149.1 2332 UTR 3 NP_001276078.1
NM_018668.4 2332 UTR 3 NP_061138.3
XM_005254887.1 2332 UTR 3 XP_005254944.1
XM_011521448.1 2332 UTR 3 XP_011519750.1
XM_011521449.2 2332 UTR 3 XP_011519751.1
XM_017022075.1 2332 UTR 3 XP_016877564.1
XM_017022076.1 2332 UTR 3 XP_016877565.1

View Full Product Details