Product Details

SNP ID
rs3739161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207766628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACAGTTCAAAGAAACCTGAAC[C/T]AAGTGGAACTTCATTACAACGCCAA
Phenotype
MIM: 601723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FZD5 PubMed Links

Gene Details

Gene
FZD5
Gene Name
frizzled class receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003468.3 2522 UTR 3 NP_003459.2

View Full Product Details