Product Details

SNP ID
rs3743110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64170264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAGGAACCAGATCCTTCACAGT[A/G]GTACCTCATATGAAAACACCATGGG
Phenotype
MIM: 606274 MIM: 123841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
There are no transcripts associated with this gene.

View Full Product Details