Product Details

SNP ID
rs3744712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5472133 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATGCAATTTGTATGTCCATTTA[C/T]TTTTAAATGAACGTATACCGTATTT
Phenotype
MIM: 610304 MIM: 614405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DERL2 PubMed Links

Gene Details

Gene
DERL2
Gene Name
derlin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304777.1 3340 UTR 3 NP_001291706.1
NM_001304779.1 3340 UTR 3 NP_001291708.1
NM_016041.4 3340 UTR 3 NP_057125.2
Gene
DHX33
Gene Name
DEAH-box helicase 33
There are no transcripts associated with this gene.

Gene
LOC105371506
Gene Name
uncharacterized LOC105371506
There are no transcripts associated with this gene.

View Full Product Details