Product Details

SNP ID
rs3746822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46174358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGAGGAGGAGGAATGGAAGAGTC[C/G]GCTCCTAGCAAGTCCCCCCTCCGTC
Phenotype
MIM: 609920
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDH22 PubMed Links

Gene Details

Gene
CDH22
Gene Name
cadherin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021248.2 3237 UTR 3 NP_067071.1
XM_011528994.2 3237 UTR 3 XP_011527296.1
XM_011528995.1 3237 UTR 3 XP_011527297.1

View Full Product Details