Product Details

SNP ID
rs3732188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47807998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTCATAGTGTCAACTGACCTTG[G/T]GTATCCATTTTTAATACAGTCTCTT
Phenotype
MIM: 607871 MIM: 600678
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO11 PubMed Links

Gene Details

Gene
FBXO11
Gene Name
F-box protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190274.1 2924 UTR 3 NP_001177203.1
NM_025133.4 2924 UTR 3 NP_079409.3
XM_005264572.4 2924 UTR 3 XP_005264629.1
XM_005264573.4 2924 UTR 3 XP_005264630.1
XM_017005015.1 2924 UTR 3 XP_016860504.1
XM_017005016.1 2924 UTR 3 XP_016860505.1
XM_017005017.1 2924 UTR 3 XP_016860506.1
Gene
MSH6
Gene Name
mutS homolog 6
There are no transcripts associated with this gene.

View Full Product Details