Product Details

SNP ID
rs3749272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132660470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACGTACCCCTCGCGGGCCCAGC[A/C]GAGCAACGTGGGGCGAAGGCGGCGG
Phenotype
MIM: 614288 MIM: 610552
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACAD11 PubMed Links

Gene Details

Gene
ACAD11
Gene Name
acyl-CoA dehydrogenase family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032169.4 974 Intron NP_115545.3
Gene
NPHP3-ACAD11
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
UBA5
Gene Name
ubiquitin like modifier activating enzyme 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320210.1 974 Intron NP_001307139.1
NM_001321238.1 974 Intron NP_001308167.1
NM_001321239.1 974 UTR 5 NP_001308168.1
NM_024818.4 974 UTR 5 NP_079094.1
NM_198329.3 974 Intron NP_938143.1

View Full Product Details