Product Details
- SNP ID
-
rs1800038
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:111079134 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCAAGGCGTCGCGCTGGCGCGGG[A/C]GGCAGAACCGCGAGAAGCGCTTCAC
- Phenotype
-
MIM: 104210
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADRA2A
PubMed Links
Gene Details
- Gene
- ADRA2A
- Gene Name
- adrenoceptor alpha 2A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000681.3 |
2103 |
Silent Mutation |
AGG,CGG |
R380R |
NP_000672.3 |
View Full Product Details