Product Details

SNP ID
rs3761248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3069747 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTCCCAGCAGCTGGGTGGCAGG[C/T]GCTTTCACAAAGGGAGAGTTGAGCA
Phenotype
MIM: 167050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OXT PubMed Links

Gene Details

Gene
OXT
Gene Name
oxytocin/neurophysin I prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000915.3 Intron NP_000906.1
XM_011529238.1 Intron XP_011527540.1

View Full Product Details