Product Details
- SNP ID
-
rs3753852
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:205047339 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- CTGAGATCCTAGCAGCCCTCACCCA[C/T]GCCACCACAGAGCCCTGTACTGACC
- Phenotype
-
MIM: 190197
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CNTN2
PubMed Links
Gene Details
- Gene
- CNTN2
- Gene Name
- contactin 2
View Full Product Details