Product Details

SNP ID
rs3760193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4557150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCCCCAGGCCCTACTGCTTTTTC[A/G]TTATTACCCTGATTTTCTTAGCTGA
Phenotype
MIM: 612341 MIM: 604885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGT6 PubMed Links

Gene Details

Gene
GGT6
Gene Name
gamma-glutamyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122890.2 2326 UTR 3 NP_001116362.1
NM_001288702.1 2326 UTR 3 NP_001275631.1
NM_001288703.1 2326 UTR 3 NP_001275632.1
NM_001288704.1 2326 UTR 3 NP_001275633.1
NM_153338.3 2326 UTR 3 NP_699169.2
Gene
MYBBP1A
Gene Name
MYB binding protein 1a
There are no transcripts associated with this gene.

View Full Product Details