Product Details
- SNP ID
-
rs3775110
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:1168234 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATGTTTCCAAAGCAAAGTGAAAAC[A/G]CCTCTTTTGCACCCCACAGGCCAAG
- Phenotype
-
MIM: 605918
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPON2
PubMed Links
Gene Details
- Gene
- SPON2
- Gene Name
- spondin 2
View Full Product Details