Product Details

SNP ID
rs3790028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50999858 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGAGGCTCAGCCGTCTATACTAT[C/G]AACAGGTTGTTCCACAGATGGTCTG
Phenotype
MIM: 603695
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GUCY1B2 PubMed Links

Gene Details

Gene
GUCY1B2
Gene Name
guanylate cyclase 1 soluble subunit beta 2 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details