Product Details

SNP ID
rs3791577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:239195219 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCCTTGGTTTACCGGACTGCCA[A/C]GTGAAACTCCACTATCTGCTAAAGT
Phenotype
MIM: 605314
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HDAC4 PubMed Links

Gene Details

Gene
HDAC4
Gene Name
histone deacetylase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006037.3 Intron NP_006028.2
XM_006712877.3 Intron XP_006712940.1
XM_006712878.3 Intron XP_006712941.1
XM_006712879.3 Intron XP_006712942.1
XM_006712880.3 Intron XP_006712943.1
XM_011512217.2 Intron XP_011510519.1
XM_011512218.2 Intron XP_011510520.1
XM_011512219.2 Intron XP_011510521.1
XM_011512220.2 Intron XP_011510522.1
XM_011512221.1 Intron XP_011510523.1
XM_011512222.2 Intron XP_011510524.1
XM_011512223.2 Intron XP_011510525.1
XM_011512224.2 Intron XP_011510526.1
XM_011512225.2 Intron XP_011510527.1
XM_011512226.2 Intron XP_011510528.1
XM_011512227.2 Intron XP_011510529.1
XM_011512230.1 Intron XP_011510532.1
XM_017005394.1 Intron XP_016860883.1
XM_017005395.1 Intron XP_016860884.1
Gene
MGC16025
Gene Name
uncharacterized LOC85009
There are no transcripts associated with this gene.

View Full Product Details