Product Details

SNP ID
rs3781236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92594106 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAATAACTTCCCAATTTTAAAAT[A/G]AGCTAGAAACTGCTGCTTAGTTAGA
Phenotype
MIM: 148760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF11 PubMed Links

Gene Details

Gene
KIF11
Gene Name
kinesin family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004523.3 Intron NP_004514.2

View Full Product Details