Product Details
- SNP ID
-
rs3199777
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:8861116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGACTCTAGACACTTGGTGGAAAG[G/T]GAGGCGAGAAAAACAATGACTTGGG
- Phenotype
-
MIM: 172430
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ENO1
PubMed Links
Gene Details
- Gene
- ENO1
- Gene Name
- enolase 1
- Gene
- MIR6728
- Gene Name
- microRNA 6728
There are no transcripts associated with this gene.
View Full Product Details