Product Details
- SNP ID
-
rs3809373
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:102801022 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACAAAATATAACCTGAAAAGACTA[C/T]AACCTAGGCGATGAAGATTGGCTTT
- Phenotype
-
MIM: 611613
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BIVM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79164513] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BIVM
- Gene Name
- basic, immunoglobulin-like variable motif containing
- Gene
- BIVM-ERCC5
- Gene Name
- BIVM-ERCC5 readthrough
There are no transcripts associated with this gene.
- Gene
- KDELC1
- Gene Name
- KDEL motif containing 1
There are no transcripts associated with this gene.
View Full Product Details