Product Details

SNP ID
rs3810365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45259360 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGATGTTCCCAGAACCACCCCC[A/G]TGGGAAAAGAGAACTAATTTCATGT
Phenotype
MIM: 606495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARK4 PubMed Links

Gene Details

Gene
MARK4
Gene Name
microtubule affinity regulating kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199867.1 Intron NP_001186796.1
NM_031417.3 Intron NP_113605.2
XM_006723307.3 Intron XP_006723370.1

View Full Product Details