Product Details

SNP ID
rs3792804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154947901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTAGAACTTAAAACAGGATAAG[C/T]ATTTCATTCAGGAGATAATAAGACG
Phenotype
MIM: 607005 MIM: 611835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GEMIN5 PubMed Links

Gene Details

Gene
GEMIN5
Gene Name
gem nuclear organelle associated protein 5
There are no transcripts associated with this gene.

Gene
MRPL22
Gene Name
mitochondrial ribosomal protein L22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014990.2 Intron NP_001014990.1
NM_014180.3 Intron NP_054899.2

View Full Product Details