Product Details

SNP ID
rs3729964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3084573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTACCTGTCTCAGCTCCAGGTC[A/G]GACATGGCCCTCTTGCCGCCCCTCG
Phenotype
MIM: 192340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AVP PubMed Links

Gene Details

Gene
AVP
Gene Name
arginine vasopressin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000490.4 152 Silent Mutation TCC,TCT S34S NP_000481.2
XM_011529267.1 152 Silent Mutation TCC,TCT S34S XP_011527569.1

View Full Product Details