Product Details

SNP ID
rs3738723
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:226633438 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGCAAATTTTACATTCCCTTCCATA[A/T]CAGACTCACTGGCTTTCAGTTAAAA
Phenotype
MIM: 147522
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITPKB PubMed Links

Gene Details

Gene
ITPKB
Gene Name
inositol-trisphosphate 3-kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002221.3 4414 UTR 3 NP_002212.3
XM_005273120.2 4414 Intron XP_005273177.1
XM_017001211.1 4414 Intron XP_016856700.1

View Full Product Details