Product Details

SNP ID
rs3803690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89696951 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGGACACGTGGAGGACCCCCAA[C/G]TCCTGAGCCCCGTACTCCGTACTGC
Phenotype
MIM: 603464
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 2162 Intron NP_001092003.2
NM_001160367.1 2162 Intron NP_001153839.1
NM_052987.3 2162 Intron NP_443713.2
NM_052988.4 2162 Intron NP_443714.3
XM_006721308.2 2162 Intron XP_006721371.1
XM_006721310.2 2162 Intron XP_006721373.1
XM_011523405.2 2162 Intron XP_011521707.1
XM_011523406.2 2162 Intron XP_011521708.1
XM_011523407.2 2162 Intron XP_011521709.1
XM_011523408.2 2162 Intron XP_011521710.1
XM_011523410.2 2162 Intron XP_011521712.1
XM_011523411.2 2162 Intron XP_011521713.1
XM_011523412.1 2162 Intron XP_011521714.1
XM_011523413.2 2162 Intron XP_011521715.1
XM_011523414.2 2162 Intron XP_011521716.1
XM_011523415.2 2162 Intron XP_011521717.1
XM_011523416.2 2162 Intron XP_011521718.1
XM_011523417.2 2162 Intron XP_011521719.1
XM_017023806.1 2162 Intron XP_016879295.1
XM_017023807.1 2162 Intron XP_016879296.1
XM_017023808.1 2162 Intron XP_016879297.1
XM_017023809.1 2162 Intron XP_016879298.1
XM_017023810.1 2162 Intron XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152339.3 2162 UTR 3 NP_689552.2
XM_005256279.4 2162 UTR 3 XP_005256336.1

View Full Product Details