Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001002266.2 | 3231 | UTR 3 | NP_001002266.1 | ||
NM_001282866.1 | 3231 | UTR 3 | NP_001269795.1 | ||
NM_145021.5 | 3231 | UTR 3 | NP_659458.2 | ||
XM_005271804.2 | 3231 | UTR 3 | XP_005271861.1 | ||
XM_006717704.2 | 3231 | UTR 3 | XP_006717767.1 | ||
XM_011539492.2 | 3231 | UTR 3 | XP_011537794.1 | ||
XM_011539493.2 | 3231 | UTR 3 | XP_011537795.1 | ||
XM_011539494.2 | 3231 | UTR 3 | XP_011537796.1 | ||
XM_011539495.1 | 3231 | UTR 3 | XP_011537797.1 | ||
XM_017015894.1 | 3231 | UTR 3 | XP_016871383.1 |