Product Details

SNP ID
rs3825057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118752276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGGTAGGCAACCCTGAGATAGA[C/T]TGGGGGCCTGAATGAATGTGAATGT
Phenotype
MIM: 600326
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX6 PubMed Links

Gene Details

Gene
DDX6
Gene Name
DEAD-box helicase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257191.2 Intron NP_001244120.1
NM_004397.5 Intron NP_004388.2
XM_005271417.3 Intron XP_005271474.1
XM_011542644.1 Intron XP_011540946.1
XM_011542645.1 Intron XP_011540947.1
XM_017017251.1 Intron XP_016872740.1

View Full Product Details