Product Details
- SNP ID
-
rs3815019
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:20904964 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTATATTTCCAGTAAGAAGTATGC[A/G]CTGTGGTCTGAAGATAAGCGGTGCT
- Phenotype
-
MIM: 616167
MIM: 614709
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DCUN1D3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115829564] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCUN1D3
- Gene Name
- defective in cullin neddylation 1 domain containing 3
There are no transcripts associated with this gene.
- Gene
- LYRM1
- Gene Name
- LYR motif containing 1
View Full Product Details