Product Details

SNP ID
rs3749193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44916633 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGAACAAAGGCACGTGGAGGG[A/G]GTGGGTGGTGAATCCACTTCTTATA
Phenotype
MIM: 600585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGM4 PubMed Links

Gene Details

Gene
TGM4
Gene Name
transglutaminase 4
There are no transcripts associated with this gene.

Gene
ZDHHC3
Gene Name
zinc finger DHHC-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135179.1 2705 UTR 3 NP_001128651.1
NM_016598.2 2705 UTR 3 NP_057682.1
XM_005265209.1 2705 UTR 3 XP_005265266.1
XM_005265212.2 2705 UTR 3 XP_005265269.1
XM_006713189.1 2705 UTR 3 XP_006713252.1
XM_006713192.2 2705 Intron XP_006713255.1
XM_006713193.2 2705 Intron XP_006713256.1
XM_006713194.3 2705 UTR 3 XP_006713257.1
XM_011533797.1 2705 UTR 3 XP_011532099.1
XM_011533800.2 2705 UTR 3 XP_011532102.1
XM_011533801.2 2705 Intron XP_011532103.1
XM_017006557.1 2705 UTR 3 XP_016862046.1
XM_017006558.1 2705 UTR 3 XP_016862047.1
XM_017006559.1 2705 UTR 3 XP_016862048.1
XM_017006560.1 2705 UTR 3 XP_016862049.1
XM_017006561.1 2705 UTR 3 XP_016862050.1
XM_017006562.1 2705 UTR 3 XP_016862051.1
XM_017006563.1 2705 Intron XP_016862052.1
XM_017006564.1 2705 Intron XP_016862053.1
XM_017006565.1 2705 UTR 3 XP_016862054.1
XM_017006566.1 2705 Intron XP_016862055.1
XM_017006567.1 2705 Intron XP_016862056.1
XM_017006568.1 2705 UTR 3 XP_016862057.1
XM_017006569.1 2705 Intron XP_016862058.1
XM_017006570.1 2705 UTR 3 XP_016862059.1
XM_017006571.1 2705 Intron XP_016862060.1

View Full Product Details