Product Details

SNP ID
rs3827937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84604358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGAGACGCAGAACCTCATAGGGA[C/G]AGGGCGGGAGCACCCTTCCAAGGTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN2 PubMed Links

Gene Details

Gene
ZSCAN2
Gene Name
zinc finger and SCAN domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007072.1 Intron NP_001007073.1
NM_017894.5 Intron NP_060364.4
NM_181877.3 Intron NP_870992.2
XM_017022392.1 Intron XP_016877881.1
XM_017022393.1 Intron XP_016877882.1

View Full Product Details