Product Details

SNP ID
rs3828973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73682169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAACATAAGGAGGAGTGCCCCA[A/G]GGGGCTGACCTTAGCGGGAGGAACG
Phenotype
MIM: 615733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC30 PubMed Links

Gene Details

Gene
DNAJC30
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032317.2 1283 UTR 3 NP_115693.2
Gene
WBSCR22
Gene Name
Williams-Beuren syndrome chromosome region 22
There are no transcripts associated with this gene.

View Full Product Details