Product Details

SNP ID
rs3751538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59137366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCTCCTCTGCCCCATGTGTCA[C/G]AGTCACGGGCACCTCAGATCTTGGT
Phenotype
MIM: 601479
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYO1E PubMed Links

Gene Details

Gene
MYO1E
Gene Name
myosin IE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004998.3 3713 UTR 3 NP_004989.2

View Full Product Details