Product Details

SNP ID
rs3751862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82148624 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTAAAATGATAATCTCTTTACA[A/C]GTAAACGTTACAGTATTAATAACTA
Phenotype
MIM: 605500
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MPHOSPH6 PubMed Links

Gene Details

Gene
MPHOSPH6
Gene Name
M-phase phosphoprotein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005792.2 753 UTR 3 NP_005783.2
XM_011522808.2 753 UTR 3 XP_011521110.1
XM_017022852.1 753 UTR 3 XP_016878341.1

View Full Product Details