Product Details

SNP ID
rs3752128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51571566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAAGTGGCGTTTGCTAATTTTAG[C/T]TGGATAATTAGCCTGTTAATTTTCC
Phenotype
MIM: 601139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF175 PubMed Links

Gene Details

Gene
ZNF175
Gene Name
zinc finger protein 175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007147.3 Intron NP_009078.1
XM_017027249.1 Intron XP_016882738.1

View Full Product Details