Product Details

SNP ID
rs3756811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24775238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACTTTGACAGCTGCAGGGCCGC[C/G]GCCTGGGTAAGGGCGAAGGGGTTGT
Phenotype
MIM: 602842
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GMNN PubMed Links

Gene Details

Gene
GMNN
Gene Name
geminin, DNA replication inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251989.1 308 Intron NP_001238918.1
NM_001251990.1 308 Intron NP_001238919.1
NM_001251991.1 308 Intron NP_001238920.1
NM_015895.4 308 UTR 5 NP_056979.1
XM_005249159.1 308 Intron XP_005249216.1
XM_011514651.2 308 Intron XP_011512953.1

View Full Product Details