Product Details

SNP ID
rs3763412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158732395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTTGTCTATAAAGGTATTGATA[C/T]GCTTCCCAAATAATCCTATATACTA
Phenotype
MIM: 616691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESYT2 PubMed Links

Gene Details

Gene
ESYT2
Gene Name
extended synaptotagmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020728.2 4560 UTR 3 NP_065779.1

View Full Product Details